Would you like to log in again?
Life/form® I.M. Injection Simulator
Supplier: NASCO HEALTHCARE INC. SE
LF00961
470119-434EA 1238.8 USD InStock
470119-434
Life/form® I.M. Injection Simulator
Models Human Biology Models
The Perfect Volunteer For Injection Training
  • Simulated landmarks for proper positioning
  • A viewing port is located on one side to visualize what they feel
  • Includes carrying case
Here is the perfect volunteer for training your beginning healthcare students to administer intramuscular injections. Always available for class demonstrations or practice by groups of any size, the Life/form® Intramuscular Injection Simulator offers both visual and tactile learning. A simulated bony structure is embedded in the torso and represents the superior end of the femur, or greater trochanter, the posterior superior and anterior superior iliac spines, and the sacrum. The bony structure provides palpable anatomical landmarks so students can identify proper injection sites. A section of the upper, outer quadrant of the left gluteal area is cut away to allow students to visualize the underlying structures. The gluteus medius and gluteus maximus muscles, sciatic nerve, and vascular structures are clearly shown. Three types of intramuscular injections can be taught and practiced on this simulator: dorsogluteal, ventrogluteal, and vastus lateralis injections. As with all Life/form® replicas and simulators, the Intramuscular Injection Simulator is extremely realistic. Skin and muscle textures, as well as bone shape and position, closely resemble a live patient. As a result, perforation of the tissue with a needle duplicates the sensation of administering an actual injection. Comes complete with teaching guide, supply of syringes, and hard carrying case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty