Would you like to log in again?
Life/form® Central Venous Cannulation Simulator
Supplier: NASCO HEALTHCARE INC. SE
W44058 LF01087
470103-406EA 274 USD InStock
470103-406 470119-454
Life/form® Central Venous Cannulation Simulator
Models Human Biology Models
Ideal Trainer Form Acls And Atls
  • Replicable muscle and bone layer
  • Landmarks exactly like a patient
  • Allows for use of a swan-ganz catheter
Designed for learning and practicing CVC techniques, the Life/form® Central Venous Cannulation Simulator is directed to the emergency medical field. ACLS and ATLS participants will find this simulator to be the ideal trainer. Anatomically accurate, palpation is exactly the same as on a patient. The sternal notch, sternocleidomastoid, clavicle, and other supporting features make this simulator a pleasure to work with. Internal features include a replaceable muscle and bone section, subclavian, internal jugular, external jugular, and carotid. Use of a Swan-Ganz catheter is possible and changing of the internal tubing is extremely easy. The neck of the simulator is positioned to the left, making landmark identity quite easy visually. The sternal notch “sinks” 1-1.5 cm when palpated. All external and internal landmarks correlate to a live patient. Modified to meet ATLS and ACLS standards, this simulator has proven itself in the emergency medical field. The consultant for this project was Forrest M. Gridley, M.A., P.A.-C. Includes fluid supply bags, replaceable skin, needles, one quart of Life/form® blood, and instruction guide in a hard carry case.
Ordering information: Fluid supply stand not included.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty