Would you like to log in again?
Arrhythmia, Heart and Breath Sounds Tutor
Supplier: AMERICAN 3B SCIENTIFIC SE
1005892 1018192 1005891 1018193 1018150 1018191 1018182 1018185 1018175 1018194 1018184 1017976 1018180 1018186 1018181 1018177 1018190 1018183 1018189 1018176 1018179 1018188 1018178
470119-506EA 177.89 USD InStock
470119-506 470119-528 470119-518 470119-504 470119-526 470119-516 470119-524 470119-502 470119-536 470119-514 470119-522 470119-500 470119-512 470119-534 470119-520 470119-498 470119-510 470119-532 470119-496 470119-530 470119-494 470119-492 470119-490
Arrhythmia, Heart and Breath Sounds Tutor
Models Human Biology Models
Flexible And Adaptable Training
  • Multiple cartridges make it easy to change sounds
  • Students all hear the same sounds for ease of identification
Combining two tutors into one, this unit simulates either heart and breath sounds or arrhythmias. A perfect tool for Medical Schools, Nursing schools, EMS Education programs or other healthcare settings where rhythm and sound recognition is critical. This unique tutor allows instructors the flexibility to change menus by changing plug-in modules. The unit has a 16-panel keypad with two built-in sounders and two external sounders. Linking additional sounders or a speaker allows access to more users. There is a volume control and two LEDs that illuminate during systole as well as an illuminated display showing the example currently playing. Arrhythmias may be viewed by connecting the three snap-on lead patient wires to your ECG-monitor. The unit comes with a 12 Volt adapter. All sound menus are sold separately, each containing 16 different sounds.
All data used in the menus are actual patient sounds. Arrhythmias and sounds of the same name are from different patients to show appearance variations.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty