Would you like to log in again?
Isometric Goniometers
Supplier: AMERICAN 3B SCIENTIFIC SE
1005903 1009013 1005900
470119-612EA 10.25 USD InStock
470119-612 470119-610 470119-600
Isometric Goniometers
Goniometers
These clear plastic goniometers permit the observation of joint's axis of motion and range of motion.
  • Calibrated in 3 scales
  • 360 degree scale
The 360º head has three scales calibrated for use with the ISOM (International Standards of Measurement) system.
Linear scale in inches and centimeters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty