Would you like to log in again?
Baseline Pinch Gauge, AMERICAN 3B SCIENTIFIC SE
Supplier: AMERICAN 3B SCIENTIFIC SE
1015294
470119-608EA 225.8 USD InStock
470119-608
Baseline Pinch Gauge, AMERICAN 3B SCIENTIFIC SE
Dynamometers
The baseline gauge measures pinch strength, with reliable and repeatable readings.
Measures tip, key and palmar pinch strength of healthy hands,
Consistent results with the published Mathiowetz studies,
Indicator remains at the subject's maximum reading until reset,
Strength readings in pounds (60 lbs.) and kilograms (27 kg),
Lightweight anodized aluminum with wrist strap and carrying case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty