Would you like to log in again?
Baseline® Clinical Tuning Forks
Supplier: AMERICAN 3B SCIENTIFIC SE
3005258 1017425 1017431 1017430 1017427 1017429 1017432 3005260 1017426 1017434
470119-660EA 10.25 USD InStock
470119-660 470119-618 470119-628 470119-614 470119-658 470119-626 470119-624 470119-622 470119-630 470119-620
Baseline® Clinical Tuning Forks
Educational Materials Biology Educational Materials Human Biology Learning Activities
For testing hearing and sensation.
  • Available Individually or in sets
  • Variable Cycle option
  • Lightweight alluminum construction
This standard tool is designed to evaluate hearing and vibratory sensation in the industry. The clinical grade set includes six forks: 128, 256, 512, 1024, 2048, and 4096 cps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty