Would you like to log in again?
Life/form® Pediatric Head Injection Trainer
Supplier: NASCO HEALTHCARE INC. SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LF00999 LF01003 1005632
470119-838EA 307.5 USD InStock
470119-838 470119-836 470119-840
Life/form® Pediatric Head Injection Trainer
Models Human Biology Models
Practice Temporal And Jugular Infusions And Injections
  • Realistically modeled on a 6 month old
  • Life like skin allows palpation for the vein
  • Included carrying case
An invaluable tool for demonstrating and practicing infusions and injections in the temporal and jugular veins of a newborn to a twelve-month-old infant. The neck is made of soft, flexible foam to provide a realistic feel of palpation and puncture. The pediatric head includes the following: Head (to represent a six-month-old infant) with skin and veins, Fluid supply bag, 2 different gauge winged infusion needles, Artificial blood.
Supplied with hard carrying case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty