Would you like to log in again?
Gaumard® Pediatric Nursing Care Mannequin
Supplier: AMERICAN 3B SCIENTIFIC SE
1005808 1005839 1005796
470119-924EA 182.69 USD InStock
470119-924 470119-328 470119-326
Gaumard® Pediatric Nursing Care Mannequin
Models Human Biology Models
High Quality Pediatric Care Simulator
  • Soft, lifelike face skin
  • Self-molded hair
  • Eyes open and close in realistic eye sockets for ophthalmic procedures
This manikin simulates a five-year old child with rotatable arms and legs (jointed elbows and knees) and articulating head.
It allows training of basic and advanced pediatric patient care procedures such as:
Bathing and bandaging
Ophthalmic exercises
Oral and dental hygiene (articulating jaw with teeth and tongue)
Tube feeding and gastric suction
I.M. injection (upper thigh and deltoid)
Tracheotomy care
Male and female catheterization
Enema administration
Supplied with T-Shirt, shorts, neck brace and carrying bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty