Would you like to log in again?
3B Scientific® Functional Physiological Skeleton
Supplier: AMERICAN 3B SCIENTIFIC SE
1020180
470120-368EA 2142.9 USD InStock
470120-368
3B Scientific® Functional Physiological Skeleton
Models Human Biology Models
Show Your Students How The Skeleton Moves
  • Half The Size Of A Normal Skeleton
  • Available With Or Without Painted Muscle Locations
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
This skeleton features an impressive new assembly of all the joints that enable it to make individual movements that are almost true-to-life. Demonstrate the complex movement sequences of the upper limb. The shoulder blades twist when the arms are raised. Even the position of spoke bones and ulna in relation to each other during internal and external rotation of the forearm can be shown clearly. The flexible spine of this skeleton can make natural movements (lateral bend, movement and stretching, rotation). When the knee joint is stretched and bent, thanks to the flexible assembly, the position of the knee cap can be demonstrated. The hand bones have been assembled on wire to show their natural position. The flexible assembly of the foot bones makes them maneuverable. Even the low level of maneuverability of the sacroiliac joint and the sacrococcygeal symphysis can be demonstrated. Now available on a stable metal hanging stand with 5 casters.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty