Would you like to log in again?
Life/form® Airway Larry Trainer
Supplier: AMERICAN DIAGNOSTIC CORP CP
LF03609 LF03605 4000 LF03699 LF03623 LF03606
470119-918EA 12.4 USD InStock
470119-918 470119-902 470119-900 470137-878 470120-422 470120-424
Life/form® Airway Larry Trainer
Models Human Biology Models
Practice Intubation, Suction, And Ventilation Techniques
  • Allow for oral, nasal, and digital intubations
  • Head can be placed onto a cparlene® manikin
  • Includes carrying case
The Life/form® Airway Management Trainer simulates non-anesthetized patients for practicing intubation, ventilation, and suction techniques. Realistic anatomy and landmarks including teeth, tongue, oral and nasal pharynx, larynx, epiglottis, arytenoids, false cords, true vocal cords, trachea, lungs, esophagus, and stomach. The trainer allows you to practice oral, digital, and nasal intubation, as well as E.T., E.O.A., P.T.L., L.M.A., E.G.T.A., and Combitube® insertion. Suction techniques and proper cuff inflation may also be practiced and evaluated. Features durable, rugged, one-piece construction and bifurcated lungs. “Airway Larry” represents an adult with more room to maneuver than the Adult Trainer (Number available upon request). With its slightly anterior position, “Airway Larry” is a great trainer for introductory as well as advanced students. It is ideal for testing purposes and Combitube® practice. Applying pressure to the cricoid cartilage changes the position of the trachea and closes the esophagus, allowing realistic practice of the Sellick maneuver. The jaw thrust technique can also be practiced. The trainer can also be attached to Nasco’s CPARLENE®, CRiSis™ Manikin. Comes with training stand to mount head, pump spray lubricant, and hand carrying hard case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty