Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CBR2™ Calculator Based Ranger System
Supplier: D&H
TICBR2 CBR2
470120-618CS 569.99 USD InStock
CA470120-618 470120-618
CBR2™ Calculator Based Ranger System
Calculators Desktop Calculators
Measure distance, velocity and acceleration on up to 200 samples per second and distances between 15 cm and 6 m.
  • Measure up to 200 samples per second
  • Compatible with many graphing calculators
The CBR2 is compatible with the TI-83 Plus and TI-84 Plus families of graphing calculators, as well as the TI-73 Explorer and the TI-Nspire handheld, using a direct USB or I/O cable connection. It also works with the TI-89T and Voyage 200. Measure distance, velocity and acceleration on up to 200 samples per second and distances between 15 cm and 6 m. Pivoting head provides versatility and convenience. This ultrasonic motion sensor is ready for your calculator without the need for the CBL system. Provides a direct cable connection to the same calculators compatible with the CBL. CBR measures motion from 0.5 to 6 meters distance. Includes ranger, cable, table clamp, manual with five experiments and four AA batteries. Purchase includes sample activities for science and math.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty