Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Vernier® Conductivity Probe
Supplier: VERNIER SOFTWARE & TECH SE
CON-BTA
470120-630EA 124.99 USD InStock
470120-630
Vernier® Conductivity Probe
Data Loggers
This probe has applications in both biology and chemistry classrooms.
  • Fast response time
  • Quick and easy calibration using our software
  • Built-in temperature compensation
  • Alternating current at its electrodes prevents polarization and electrolysis
  • Epoxy-body graphite electrode eliminates worry about corrosion of metal electrodes
Biology teachers can use this probe to demonstrate diffusion of ions through membranes or to monitor changes in ion levels in aquatic systems. Chemistry students can use it to investigate the difference between ionic and molecular compounds, strong and weak acids, or ionic compounds that yield different ratios of ions. The conductivity probe can also monitor concentration or conductivity at three different sensitivity settings. Probe takes readings in units of conductivity (μS/cm) or concentration (mg/L TDS as NaCl). Alternating current is used at its electrodes to prevent polarization and electrolysis, so that solutions being tested are not fouled. Corrosion of metal electrodes is not a problem with this epoxy-body graphite electrode.
It has built-in temperature compensation, which allows you to do your calibrations in the lab, and then make measurements outdoors without temperature changes affecting conductivity readings. The probe also has a fast response time, reaching 98% of full value in less than five seconds. You can load saved calibrations using any of the Vernier data-collection programs. You can also do a quick two-point calibration: the probe is left out of solution for one calibration point (0 μS/cm) and is placed in a known standard (1000 μS/cm solution is provided) for a second calibration point.
Operating temperature range: 0 to 80 °C
Cell constant: 1 cm⁻¹
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty