Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Chloride Ion-Selective Electrode

Supplier: VERNIER SOFTWARE & TECH SE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CL-BTA
470120-636EA 298.99 USD InStock
470120-636
Vernier® Chloride Ion-Selective Electrode
Electrodes ISE Electrodes
Measure the concentration of chloride in aqueous samples.

  • Measure salt in water or in foods

This ion gives a quick measurement of salinity of water samples. It can even measure chloride levels in ocean saltwater or salt in food samples. Electrodes are combination style, non-refillable, and gel-filled
Included with each ISE are two calibration standards and a Short-Term Soaking Bottle
Interfering ions: CN–, Br–, I–, S2-
Electrode slope (log voltage vs. concentration): –56 mV/decade
Electrode resistance: 1 to 4 MΩ
Minimum immersion: 1.1 inch

For educational use only.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty