Would you like to log in again?
Vernier® Nitrate Ion-Selective Electrode
Supplier: VERNIER SOFTWARE & TECH SE
NO3-BTA NO3-MOD
470120-638EA 298.99 USD InStock
470120-638 470219-308
Vernier® Nitrate Ion-Selective Electrode
Electrodes ISE Electrodes
Measure the concentration of nitrate in aqueous samples.
  • The Nitrate ISE has a combination-style, non-refillable, gel-filled electrode
Nitrate concentration, which can be increased by acidic rainfall, fertilizer runoff from fields, and plant or animal decay or waste, is an important parameter in nearly all water quality studies. Use the Nitrate ISE to determine the concentration of the nitrate ion in a water sample.
Like all other PVC ISE membranes, the membrane on the ISE has a limited life expectancy. However, the replaceable module of ISE allows you to simply discard the used membrane module, and replace it with a new one.
Note: We recommend that you do not purchase ISE replacement modules too far in advance of their expected time of use as degradation occurs while they are stored on the shelf.
Important: Ion-Selective Electrodes require good chemical technique and careful calibration to obtain accurate results; they are not recommended for middle school or elementary students.
For educational use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty