Would you like to log in again?
Vernier Drop Counter
Supplier: VERNIER SOFTWARE & TECH SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vernier
VDC-BTD
470120-642EA 108.99 USD InStock
470120-642
Vernier Drop Counter
Electrodes Other Electrodes
Perform accurate, automatic titrations by counting drops of titrant.
  • Wired connection with BTA plug
  • Adjustable clamp, fits most laboratory ring stands and lattices
  • Wide drop-detecting area
  • Large sensor slot, fits most conventional stick-style sensors
  • Smaller, adjustable slot for temperature sensors
  • Accurate drop counting at rates up to 6 drops/second
The drop counter precisely records the number of drops of titrant added during a titration, which is automatically converted into volume. It may be used in conjunction with other sensors, such as a pH Sensor, Conductivity Probe, or ORP Sensor to perform conductometric or potentiometric titrations.
Ordering information: LabQuest® (2, 3, Mini) interface is required but not included.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty