Would you like to log in again?
Vernier® Light Sensor
Supplier: VERNIER SOFTWARE & TECH SE
LS-BTA
470120-692EA 68.99 USD InStock
470120-692
Vernier® Light Sensor
Light Meters
Perfect for studying a wide variety of concepts.
  • For inverse square law experiments or for studying polarizers, reflectivity, or solar energy
  • Perform inverse square light intensity experiments using a point source of light and reflectivity studies
  • Demonstrate the flicker of fluorescent lamps and other lamps
  • Carry out solar energy studies and conduct polarized filter studies
  • Study light intensity in various parts of a house or school
  • Use it as part of a study of plant growth to measure light intensity
The Light sensor approximates the human eye in spectral response and can be used over three different illumination ranges, which you select with a switch.
For educational use only.
Wavelengths: 400 to 800 nm
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty