Would you like to log in again?
Vernier® Go! Link
Supplier: VERNIER SOFTWARE & TECH SE
GO-LINK GL-TP
470120-708EA 74.99 USD InStock
470120-708 470112-948
Vernier® Go! Link
Cables
Use your BTA Vernier probes with any computer
  • Max. sample capacity is 200 samples per second (computer) and 10 samples per second (chromebook)
  • 12-bit resolution:
The single-channel interface connects any Vernier sensor to your Windows or Macintosh computer. Just plug the sensor into the Go! Link, connect the Go! Link to the computer’s USB port, and begin collecting data. It comes with the highly-capable Logger Lite software for graphing and analysis of collected data, and a user’s guide.
Ordering information: The Teacher Pack contains 8 Go! Link connectors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty