Would you like to log in again?
Mechanical Engineering Models
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH0366TM-39 PH0366TM-5 PH0366TM-26 PH0366TM-23 PH0366-TM-6
470120-726EA 52.3 USD InStock
470120-726 470120-728 470120-722 470120-724 470120-720
Mechanical Engineering Models
Educational Materials Physics Educational Materials Mechanics Learning Activities
Clearly Observe Inner Workings
  • Demonstrate the most fundamental and most frequently used mechanisms with these mechanical engineering models
  • All components are precisely designed and can be assembled with clear sides to facilitate teaching
Useful for introducing engineering concepts, with models that are easy to assemble and disassemble. These models are great to visually demonstrate how the internal mechanisms of car transmissions, snowmobiles, elevators, and other mechanical items work. Students will be able to see things from how a simple transmission works with three forward gears and one reverse to the typical cone clutch drive used in many snowmobiles.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty