Would you like to log in again?
PCR-based VNTR Human DNA Typing Kit
Supplier: Edvotek
334-B
470120-892EA 129 USD InStock
470120-892
PCR-based VNTR Human DNA Typing Kit
Educational Materials Biology Educational Materials PCR Learning Activities
DNA polymorphisms are regions of DNA that vary greatly (are polymorphic) between individuals. These polymorphisms are frequently used to obtain a 'DNA fingerprint of individuals.
One polymorphic DNA region, known as the D1S80 locus variably contains between 14 and 40 copies of a 16 base pair repeat; this is an example of a VNTR (variable number of tandem repeats) polymorphism.
In this experiment, students isolate their DNA from cheek cells and perform PCR to fingerprint their DNA at the D1S80 locus, using technology used by FBI laboratories in a safe classroom experiment. Students can also quantitatively measure the size of PCR products to determine individual fingerprints. Safe experimental design enables students to perform the experiment using hair or sputum. Includes instructions and materials for 25 reactions.
Requires electrophoresis equipment, thermal cycler, white light or UV transilluminator, micropipets and microcentrifuge, all available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty