Would you like to log in again?
PCR-Based DNA Fingerprinting Kit
Supplier: Edvotek
371
470120-896EA 259.3 USD InStock
470120-896
PCR-Based DNA Fingerprinting Kit
Educational Materials Biology Educational Materials PCR Learning Activities
Students explore Forensic Science with the PCR-Based DNA Fingerprinting Kit.
  • Lab Encourages Independent Inquiry
  • Time Required: 75 Minutes plus Two Hours for PCR
  • Students Practice PCR in the Classroom
  • Instructions Included
  • PCR EdvoBeads Included
This kit provides easy-to-follow instructions, so students can develop various crime scene scenarios independently.
Plasmid DNA, when amplified by PCR, provides materials that represent individual DNA profiles. Students use these outcomes to solve a crime.
Ordering information: This kit includes instructions, PCR EdvoBeads, DNA templates, primers, DNA ladder, ultrapure water, wax beads, agarose, loading dye, electrophoresis buffer, InstaStain Ethidium Bromide and FlashBlue stain. Micropipettes that measure between 5 and 50 µl, tips, a thermal cycler, electrophoresis apparatus, a power supply, a microwave or hot plate, and a UV transilluminator are required but not included.
Packaging: Some components require freezer storage on arrival.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty