Would you like to log in again?
Determination of Protein Molecular Weight Kit (EDVO-Kit 153)
Supplier: Edvotek
153-B
470120-898EA 133.69 USD InStock
470120-898
Determination of Protein Molecular Weight Kit (EDVO-Kit 153)
Educational Materials Biology Educational Materials PCR Learning Activities
Using pre-stained LyphoProteins™, analysis with denaturing SDS vertical polyacrylamide gel electrophoresis determines subunit molecular weights.
  • Material for Six Lab Groups Sharing Three Gels
  • Lab Uses Pre-Stained LyphoProteins
  • Vertical Electrophoresis Apparatus Required
Pre-stained proteins with unknown molecular weights are assigned weights based on the relative mobility of pre-stained standard protein markers.
The kit includes material for six lab groups sharing three gels and requires a vertical electrophoresis apparatus, power supply, micropipets, and polyacrylamide gels, available separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty