Would you like to log in again?
Simulation of HIV Detection by Western Blot Kit
Supplier: Edvotek
275
470120-908EA 175.15 USD InStock
470120-908
Simulation of HIV Detection by Western Blot Kit
Blotting Kits
This kit provides a hands-on examination of immunology procedures.
  • Students Practice Immunology Procedures
  • Time Required: ~75 min over Two Days
  • Instructions Included
The second assay used to confirm a positive HIV ELISA result is the Western Blot.
Students separate protein samples from hypothetical patients on agarose gels, transfer the samples to a membrane, and detect the simulated HIV proteins.
Ordering information: This kit includes instructions, samples, standard molecular weight markers, protein agarose, various buffers and reagents, PVDF membrane, filter paper, stain, 1 mL pipette, and an 100 mL graduated cylinder. Electrophoresis apparatus, power supply, micropipettes with tips, a microwave or hot plate, an incubation oven, a shaker platform, lab glassware, small plastic trays, micro test tubes, pipette pumps or bulbs, metric rulers, distilled water, methanol, and glacial acetic acid are required but not included.
Delivery information: Some components require refrigerator storage on arrival
Packaging: Some components require refrigerator storage on arrival
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty