Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PCR Based Identification of Foodstuffs for Genetically Modified Organisms

Supplier: Edvotek
962-B
470120-912EA 258.3 USD InStock
470120-912
PCR Based Identification of Foodstuffs for Genetically Modified Organisms
Educational Materials Biology Educational Materials PCR Learning Activities
This activity works in conjunction with AP Biology classes.

  • Students Test for Genetic Engineering
  • Time Required: 75 Minutes plus Two Hours for PCR
  • Activity in Conjunction with AP Biology
  • Application: Extracting DNA from Food or Plants

Some foods contain raw materials from genetically modified organisms (GMO). In this experiment, students extract DNA from food or plant material and perform PCR to determine if any GMO indicator genes are present.

Amplified DNA is separated and sized by agarose gel electrophoresis.

Ordering information: This item includes instructions, DNA extraction reagents, PCR EdvoBeads, microtubes, primers, a DNA ladder, ultrapure water, wax beads, gel loading dye, agarose, electrophoresis buffer, InstaStain Ethidium Bromide, and FlashBlue stain. Micropipettes to measure between 5 and 50 µl, tips, water bath, microcentrifuge, a thermal cycler, electrophoresis apparatus, a power supply, a microwave or hot plate, and a UV transilluminator are required but not included.

Delivery information: Some components require freezer storage on arrival. This item requires shipping in one to three days.

Packaging: Some components require freezer storage on arrival

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty