Would you like to log in again?
Mega Molecules — The Structure of Water
Supplier: Mega Molecules
BC17
470120-982EA 14.7 USD InStock
470120-982
Mega Molecules — The Structure of Water
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Construct Molecules with Magnetic Parts
  • Students explore the attractions between atoms and molecules that are responsible for the physical properties of liquids and solids
Kit includes two magnetic water molecules with the hydrogen molecule attracted to the lone pair of electrons on the other water molecule. The oxygen atom attracts the electrons within the covalent bonds leaving the hydrogen atoms with a slightly positive charge. You can download free student activity sheets for this kit at www.megamolecules.com.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty