Would you like to log in again?
Hand Centrifuge
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CENTFG1
470121-084EA 131 USD InStock
470121-084
Hand Centrifuge
Centrifuges Benchtop Centrifuges
This hand-driven centrifuge is ideal for simple classroom demonstrations.
  • Easy grip handle
  • Holds four tubes at once
The centrifuge features a painted metal body and head that can hold up to four tubes, which are included, and comes with a table-mounting clamp. The three-piece construction makes for easy disassembly and storage in its molded foam box.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty