Would you like to log in again?
Hall Effect Apparatus
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SEB01 SK006
470121-160EA 1143.3 USD InStock
470121-160 470227-534
Hall Effect Apparatus
Educational Materials Physics Educational Materials Quantum Mechanics Learning Activities
This apparatus determines hall voltage and coefficient.
  • Heavy-Duty Electromagnet
  • Gauss Meter with a Thin Probe
  • Manual
  • Semiconductor Ge Sample
  • Digital Multimeter
The most distinguishing feature allows the student to individually mount a Ge crystal.
Students can also determine the type of charge carrier, the charge density of carriers, and the resistivity of a given sample.
Ordering information: The unit includes a Gauss meter with probes; digital multimeter; semiconductor 'Ge' sample, p type; and Hall effect apparatus. A 12V power supply is required but not provided.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty