Would you like to log in again?
CENCO® Photoelectric Effect Apparatus
Supplier: VWR International
PEEA01
470121-248EA 4060.15 USD InStock
470121-248
CENCO® Photoelectric Effect Apparatus
Educational Materials Physics Educational Materials Quantum Mechanics Learning Activities
Precise Determinations of Photoelectric Effect
  • Study electron photoemission from a metal surface
  • Obtain the characteristic curves of a vacuum photocell
  • Explore the effect of illumination intensity using the inverse square law
Examine the photocurrent produced by an illuminated cathode with this convenient apparatus. A current-controlled filament lamp in a light tight box illuminates a photocell with a mixed-alkali cathode. The control unit supplies a controlled bias voltage and delivers the photocurrent to a sensitive current amplifier so that the characteristic curve ofthe photocell as a function ofthe bias voltage can be precisely measured. The filament lamp is mounted on a slide with a scale, so that the relative illumination level can be adjusted by changing the distance of the lamp from the photocell. This enables a characteristic curve of photocurrent as a function of illumination to be constructed. The control unit carries multi-tum potentiometers for adjusting the lamp current and the bias voltage, and pushbuttons for selecting the photocurrent measuring range. The lamp current, bias voltage, and photocurrent are displayed on three built-in digital readouts. A user manual is included, in addition to a test box and set of connecting cables. 110 VAC, 128 W. Size: Test Box: 15cm H x 13cm W x 62cm L; weight 3.6 kg. Control Unit: 10cm H x 35cm W x 28cm D; weight 4.4 kg.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty