Would you like to log in again?
Four Point Probe Experiment
Supplier: SILICON VALLEY SCIENCE LABS SE
DFP-RES
470121-458EA 5664.15 USD InStock
470121-458
Four Point Probe Experiment
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Quick Measurements Within or Amongst Samples
Measure a wide range of resistivities at various temperatures with a high degree of accuracy. The widely-used standard of the four probe method is made even more accurate with this experiment. The collinear arrangement of these four probes removes error due to contact resistance in the electrical measurement of semiconductors by the use of two extra contacts (probes) between the current contacts. The contact resistance may be high compared to the sample resistance, but as long as the resistance of the sample and contact resistances are small compared with the effective resistance of the voltage measuring device (potentiometer, electrometer or electronic voltmeter), the measured value will remain unaffected. Because of the use of pressure contacts, this arrangement is specifically useful for quick measurements on different samples or sampling different parts of a sample. Everything required for measurements is provided, including the four probes arrangement, PID controlled oven, constant current source, and DC microvoltmeter. The constant current source provided features CCS-01 for low to medium resistivity samples and LCS-02 for high resistivity samples. 110/220 V, 50 Hz/60 Hz.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty