Would you like to log in again?
Digital Millikan Apparatus, One Piece
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SV038 SV040
470121-482EA 1989.7 USD InStock
470121-482 470159-598
Digital Millikan Apparatus, One Piece
Educational Materials Physics Educational Materials Modern Physics Learning Activities
This Digital Millikan Apparatus provides clear visibility of spheres and conducts determinations in two different ways.
  • Precision Microscope Optics
  • Easy-To-Adjust Voltage Setting
  • Polarity-Reserving Switch
  • Potentiometer
  • Voltmeter
Users can allow a selected sphere to fall through a measured distance with no electric field present then apply an electric field and observe the velocity of the rising sphere. They could also allow a sphere to fall freely then apply a high-voltage field until the sphere is suspended. Built-in to this compact tabletop unit are all the accessories you need to perform Millikan experiments, including a microscope, an illuminator, a 0—500VDC voltmeter, and all the necessary power inputs. Pure olive oil is recommended, but not included.
Once it's on a lab table and plugged into a 110VAC outlet, it is ready to use. For added convenience, the spray bottle system features a reservoir that attaches to the top of the unit, so you don’t have to look away during the experiment. Controls include an on-off switch, a polarity-reversing switch, a potentiometer for fine control of plate voltages, and a voltmeter for indicating plate voltage. A digital voltmeter also provides easy and accurate readings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty