Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Millikan Apparatus

Supplier: THE SCIENCE SOURCE SE
16120
470121-618EA 1101.05 USD InStock
470121-618
Millikan Apparatus
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Assembles In Seconds

Determine accurately the elementary charge of the electron and demonstrate quantization with this classic apparatus. It features a whole–plate reversible polarity electrode chamber, which provides a completely uniform field, and a high intensity LED light source, which is fully adjustable for background contrast and beam focus, to illuminates particles brilliantly. The focus can be adjusted to either coarse or fine through the short depth–of–field viewing scope with reticle and individual particles of the particle cloud may also be scanned. Includes the base unit with integrated 500V supply, high intensity LED light block, parallel whole–plate field chamber, viewing scope with reticle, focusing rod, atomizer, latex sphere suspension and an experimental manual.

Specifications:
•Field voltage: 0-500V; reversible polarity
•Light source: 1A high intensity LED;heat-managed
•Particle: 1 micrometer latex, in suspension
•Supply input: 110VAC 50-60Hz

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty