Would you like to log in again?
Basic Chromatography Kit
Supplier: LAB-AIDS INC (KITS) SE
15
470121-696EA 176.2 USD InStock
470121-696
Basic Chromatography Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
This kit is a exceptional introduction to chromatography.
  • Kit Shows How Chromatography Identifies Makeup of Organic and Inorganic Substances
  • Students Compare Effects of Different Solvents
  • Required Materials Included
  • Chromatography Tubes and Holders Included
  • Dyes Included
Students learn how chromatography can identify the makeup of organic and inorganic substances.
They perform experiments to compare the effects of different solvents on the separation of chlorophyll extract and dyes into their constituent components.
Ordering information: This kit includes chromatography tubes and holders, chromatography strips, developers I, II, III, and IV, capillary applicators, chloroplast extract, and mixed dyes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty