Would you like to log in again?
Lab 1: Diffusion and Osmosis
Supplier: Edvotek
281
470121-744KT 170.85 USD InStock
470121-744
Lab 1: Diffusion and Osmosis
Educational Materials Biology Educational Materials AP Biology Learning Activities
The selective permeability of membranes is of fundamental importance to cellular physiology. Students will perform experimental procedures involving diffusion, dialysis, water potential and osmotic potential. For 10 lab groups.
Duration: 60-90 minutes or two lab periods.
Kit contents:
• Sample A: orange dye
• Sample B: violet dye
• M Sucrose/1 M NaCl solution
• Powdered sucrose
• Dialysis tubing
• Instructions
Required, but not included: beakers, 1, 5 and 10 ml pipets, thermometers, scales, cork borers or small kitchen knives, graph paper, distilled or deionized H₂O, potatoes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty