Would you like to log in again?
Extension Clamp with 3-Finger Jaws
Supplier: United Scientific Supplies
ECLJ03
470121-782EA 68.8 USD InStock
470121-782
Extension Clamp with 3-Finger Jaws
Clamps General Purpose Clamps
Firmly hold glassware in place on a support stand.
  • Jaws are coated with rubber
  • Chrome plated steel rod
Features a 3-prong die-cast clamp attached to a chrome-plated steel rod. Prongs are covered with rubber sleeves. Overall length is 10".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty