Would you like to log in again?
Erada-Stain Biological/Chemical Stain Remover
Supplier: Cambridge
Erada-Stain
206
470121-812EA 39.95 USD InStock
470121-812
Erada-Stain Biological/Chemical Stain Remover
Cleaning Agents Detergents and Surface Cleansers
Erada-Stain® is a very mild cream cleaner / stain remover that can be used on hands or lab equipment.
Rapidly removes Wright’s stain and most medical and histological stains from hands, glassware and laboratory equipment. Stains disappear in seconds when applied. Rinse with water. Cream is available in 170 g squeeze tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty