Would you like to log in again?
Steel Crucibles
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SSR020-L SSR030 SSR030-L SSR020
470121-880EA 7.9 USD InStock
470121-880 470121-874 470121-882 470121-876
Steel Crucibles
Crucibles
Excellent Corrosion Resistance.
  • Constructed of 1-piece steel sheet
  • Uniform wall thickness
Unibody construction from Type 304 stainless steel. Can withstand temperatures from –180 to +800 °C. Covers included.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty