Would you like to log in again?
Hoffman Electrolysis Apparatus
Supplier: MONTCLARE SCIENTIFIC GLASS SE
SPECIAL
470122-032EA 33.39 USD InStock
470122-032
Hoffman Electrolysis Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Classic Design for Quantitative Work
Maximum convenience and minimum problems with breakage are assured with this large, eye-catching Hoffman form electrolysis apparatus. Designed for precise quantitative measurements, the two large 60mL graduated gas collecting tubes feature easy-to-read 0.2mL divisions. Stopcocks with ground glass connections retain or release gases with a twist of a finger. Glass components are mounted on an acid-resistant plastic panel; the reservoir (filling tube) is recessed into the panel for added protection. Convenient screw cap binding posts at the bottom make connections a cinch. The apparatus comes complete with a sturdy support stand and clamp, graduated H tube with stopcocks, reservoir tube, back panel and glass platinum electrodes (not shown). Overall height is 65cm. With instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty