Would you like to log in again?
High-Frequency Tesla Coil
Supplier: ELECTRO-TECHNIC PRODUCTS SE
11031 11011
470005-396EA 255.95 USD InStock
470005-396 470122-120
High-Frequency Tesla Coil
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The high-frequency coil can be used to test vacuum equipment for leakage or excite electrical tubes for demonstrations.
  • Coil tests vacuum equipment for leakage or excites electrical tubes for demonstrations
  • Instructions included
  • Connecting cord and attachment plug included
  • Coil is mounted in Bakelite case
  • Small enough to hold in one hand
It's mounted in a hard Bakelite case small enough to hold in one hand. One terminal is attached to the pointed end of the coil. A screw at the large end is adjustable.
The voltage is 40 to 50 kV, and the spark frequency is three to four MHz. A featured knob adjusts the spark length. After 10 minutes of continuous use, the coil must be turned off to cool.
Ordering information: This coil includes a connecting cord, attachment plug, and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty