Would you like to log in again?
CPEP Nuclear Science Charts
Supplier: CONTEMPORARY PHYSICS EDUCATION
296231 296200 296241 296212
470073-046EA 25.25 USD InStock
470073-046 470072-810 470122-542 470076-274
CPEP Nuclear Science Charts
Charts
Visualize nuclear science.
  • Describes basic nuclear processes and the modern picture of the nucleus and then applies them to the formation of the Universe
Nuclear Science, part of the highly popular Contemporary Physics Education Project (CPEP) series, explores the structure, properties and interactions of atomic nuclei.
It illustrates the known isotopes with a chart of the nuclides in graphic form, traces radioactive decay from alpha to gamma and shows how new elements are found. Many of the applications of nuclear science are described.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty