Would you like to log in again?
Bull's Eye Level
Supplier: GEIER & BLUHM INC SE
2-12000
470122-634EA 89 USD InStock
470122-634
Bull's Eye Level
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Perfect for leveling ripple tanks, before filling with water, this circular spirit level is enclosed in a chrome plated brass case.
Ø: 3 cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty