Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Doublet Magnifier

Supplier: United Scientific Supplies
MPF010
470122-662EA 24.7 USD InStock
470122-662
Doublet Magnifier
Magnifiers
This compact, highly corrected magnifier comes in a convenient folding holder and case.

  • Diameter: 1.3 cm
  • Focal Length: 2.7 cm
  • Magnification: 10x
  • Convenient Folding Holder and Case
  • Highly Corrected

It is 1.3 cm in diameter and has a focal length of 2.7 cm.

The magnification is 10x.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty