Would you like to log in again?
Precision Test Strips
Supplier: PRECISION LABORATORIES SE
NAT-1V-50
470123-128PK 15.9 USD InStock
470123-128
Precision Test Strips
Environmental Test Kits
Precision Test Strips quickly test for low levels of nitrite and chlorine.
  • Fast Tests
  • Pool Test Strip Measures pH, free, and Total Chlorine
  • Aquarium Test Strip Measures pH and Chlorine Levels
  • Fifty Test Strips in One Package
  • Test Strips for Nitrite and Chlorine
After immersing the strip in the solution to be measured, the concentration is measured by comparing the strip’s color to a color chart on the package.
The Pool Test Strip is three tests in one—pH, free, and total chlorine. The aquarium test strip simultaneously measures pH and chlorine levels.
Packaging: Each package includes 50 test strips in a resealable package.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty