Would you like to log in again?
Absorber Set
Supplier: SPECTRUM TECHNIQUES, LLC
RAS20
470123-270KT 481 USD InStock
470123-270
Absorber Set
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Perfectly Suited for Use with Our Nuclear Labs.
  • Complete set for radiation studies
  • Includes storage case
This set of 20 calibrated radiation absorbers covers the range from 4.5 to 7400 mg/cm2 and consists of aluminum, plastic and lead plates. The absorber are 2.75 in2 and are designed to fit all Spectrum Techniques detector stand assemblies.
The set is suitable for absorption studies with Alpha, Beta and Gamma radiation.
Delivery information: This item is Item only available to US Customers.
Packaging: One set contains twenty items.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty