Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Assortment of One-Hole Rubber Stoppers

Supplier: United Scientific Supplies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

RSTPK3
470123-294LB 20 USD InStock
470123-294
Assortment of One-Hole Rubber Stoppers
Stoppers Rubber Stoppers
One Pound of One-Hole Rubber Stoppers in Various Sizes.

  • Superior grade of rubber
  • Soft, resilent, and durable
  • Low sulfur content
  • Size numbers molded onto the top
  • All stoppers are 25 mm long
  • Sizes: #00 to #7

Designed to stay pliable over a long period of time, these stoppers can withstand temperatures from –25 to 70°C (–13 to 158°F). Ideal for use with nonaggressive or concentrated solutions.

Certifications: CAUTION: Item contains latex, which is known to cause allergic reactions in some people. Use with caution.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty