Would you like to log in again?
Direct Vision Spectroscope, Advanced
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH0594A
470123-418EA 46.59 USD InStock
470123-418
Direct Vision Spectroscope, Advanced
Educational Materials Physics Educational Materials Optics Learning Activities
Fine hand instrument for qualitative visible spectrum analysis.
  • Features adjustable slit width and draw-out focusing collar
  • Uses 3-element prisms in lieu of diffraction grating
  • Comes in an attractive and robust wooden case
Handy spectroscope for viewing the visible spectra of light sources. Wavelength dispersion is accomplished by a fixed prism rather than a diffraction grating, so the spectrum may be viewed directly rather than at an oblique angle. With an adjustable collar, the metal tube may be drawn out to focus. An adjustable light slit is fitted with achromatic objective glass and 3 element prisms. Comes complete in a wooden case for safe storage.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty