Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Steel Rods
Supplier: VWR International
WLS78306-20C WLS78306-20D S78306-20A WLS78306-20B
470123-468EA 24.45 USD InStock
470123-468 470123-466 470123-464 470123-462
Steel Rods
Support Rods and Frames
Made of zinc plated steel; designed to fit T-Nesting Support Bases.
One end threaded, with shoulder, to fit base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty