Would you like to log in again?
Round Test Tube Rack
Supplier: United Scientific Supplies
TTHP05
470123-536EA 14 USD InStock
470123-536
Round Test Tube Rack
Racks Tube Racks
Hold a variety of test tubes up to 20 mm.
  • Compact round design
  • Shipped pre-assembled
Constructed of solid hi-impact polypropylene, this preassembled rack holds 24 tubes up to 20 mm in diameter. Rack itself is 145 mm in diameter and 70 mm high. Features a perforated bottom.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty