Would you like to log in again?
DNA Fingerprinting After PCR Amplification
Supplier: Edvotek
130
470126-638KT 138.7 USD InStock
470126-638
DNA Fingerprinting After PCR Amplification
Educational Materials Forensics Educational Materials Fingerprint Analysis Learning Activities
Apply Biotechnology to Forensics Investigations
  • Convenient Ready-To-Load samples
  • Inquiry-based experiment
  • No PCR reagents are included or required
  • Time required: ~45 minutes
Forensic DNA fingerprinting is quickly becoming a widely-accepted crime fighting tool, practiced by today's criminal investigation professionals. This inquiry-based experiment, based on a crime scene scenario, includes all the supplies necessary to analyze DNA extracted and already PCR amplified from fingerprints using gel electrophoresis. No PCR reagents are included or required. Ready-to-Load™ DNA is included for quick and easy sample loading.
Kit includes: instructions, Ready-to-Load™ QuickStrip™ DNA samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain® Blue and FlashBlue™ stain, calibrated pipette, and microtipped transfer pipettes.
Required, but not included: electrophoresis apparatus, power supply, automatic micropipette and tips, balance, microwave or hot plate, visualization system.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty