Would you like to log in again?
In Search of the Cancer Gene
Supplier: Edvotek
314
470126-658KT 206.55 USD InStock
470126-658
In Search of the Cancer Gene
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Determine Genetic Predisposition for Cancer
  • Convenient Ready-To-Load samples
  • Connection to cancer genetics
  • Inquiry-based experiment
  • Time required: ~60 minutes
Suppressor genes such as p53 are essential for cell functions. Mutations in the p53 gene can be correlated to predisposition for certain cancers. Mutations in genes can either be inherited or accumulated due to environmental insults. This experiment deals with a family pedigree determination of several generations relating to cancer formation due to p53 gene mutation. This experiment does not contain human DNA.
Kit includes: instructions, Ready-to-load™ Predigested DNA samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain® Ethidium Bromide, pipette, 5 autoradiograms.
Required, but not included: electrophoresis apparatus & power supply, automatic micropipette with tips, balance, microwave or hot plate, water bath (65°C), UV Transilluminator, pipette pump or bulb, 250 ml Flasks, distilled or deionized water.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty