Would you like to log in again?
Rare Earth Magnet
Supplier: PICOSOLUTIONS GROUP INC SE
PS-2016 PS-2016
470326-954PK 7.69 USD InStock
470326-954 470126-945
Rare Earth Magnet
Magnets
Compatible with Variety of Windmill Kits
  • Strength 0.395 Tesla/3,950 Gauss
  • Size 1.875×0.875×0.375" thick
  • Pole marked with adhesive sticker
This ceramic, grade 5 magnet is ideal for use in constructing the alternator used in a variety of wind turbine kits. Use caution: ceramic magnets are magnetically strong, but physically fragile and can chip or crack easily.
Caution: WARNING! These magnets are strong and can pinch your fingers if not handled carefully. Persons wearing pacemakers should not handle strong magnets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty