Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Frog Skeleton

Supplier: ED SPELDY EAST CO. LTD.
TE37
470127-678EA 72.85 USD InStock
470127-678
Frog Skeleton
Specimens Preserved Specimens Display Specimens
This specimen block has a real toad skeleton encased.

  • Preserved and Encased in Clear Resin Material
  • Box and Information Sheet Provided
  • Length: 3"
  • Width: 3"
  • Height: 1"

The specimen is preserved and encased in clear resin material

This product comes with a box and a information sheet. The size of the acrylic is 1 x 3 x 3".

It can be observed from every side to see details.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty